Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318245.1 | NP_001305174.1 | 6 | 1895 | ||
NM_032608.6 | NP_115997.5 | 6 | 1895 | |||
XM_011530458.2 | XP_011528760.1 | 7 | 2031 | |||
XM_011530459.2 | XP_011528761.1 | 7 | 2002 | |||
XM_011530460.2 | XP_011528762.1 | 6 | 1875 | |||
XM_011530461.2 | XP_011528763.1 | 7 | 2031 | |||
XM_011530464.2 | XP_011528766.1 | 7 | 2031 | |||
XM_011530465.2 | XP_011528767.1 | 6 | 1788 | |||
XM_011530466.2 | XP_011528768.1 | 7 | 2032 | |||
XM_017029012.1 | XP_016884501.1 | 7 | 2031 | |||
XM_017029013.1 | XP_016884502.1 | 7 | 2079 | |||
XM_017029014.1 | XP_016884503.1 | 7 | 2031 | |||
XM_017029015.1 | XP_016884504.1 | 6 | 1905 | |||
XM_017029017.1 | XP_016884506.1 | 6 | 1904 | |||
GenBank mRNA | AB042648.2 | - | 2 | 207 | ||
AB075376.1 | - | 6 | 1895 | |||
AJ310931.2 | - | 6 | 1886 |