Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002581.4 | NP_002572.2 | 10 | 3389 | ||
XM_006717129.2 | XP_006717192.1 | 8 | 1463 | |||
XM_017014784.1 | XP_016870273.1 | 10 | 3429 | |||
GenBank mRNA | AK300128.1 | - | 8 | 1616 | ||
AK300129.1 | - | 4 | 320 | |||
BC078657.1 | - | 10 | 3019 | 30340336 | ||
BU158714.1 | - | 4 | 508 | |||
X68280.1 | - | 10 | 2780 |