Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: AGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000029.3 | NP_000020.1 | 3 | 1602 | ||
GenBank mRNA | AB209769.1 | - | 3 | 1114 | ||
AK222798.1 | - | 3 | 1133 | |||
AK222815.1 | - | 3 | 1134 | |||
AK293507.1 | - | 4 | 887 | |||
AK303755.1 | - | 4 | 1075 | |||
AK307978.1 | - | 4 | 1048 | |||
AK312291.1 | - | 3 | 1212 | |||
BC011519.1 | - | 3 | 1122 | 4213559 | ||
BT006851.1 | - | 2 | 1094 | |||
K02215.1 | - | 3 | 1133 | |||
W01494.1 | - | 1 | 77 |