Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201325.1 | NP_001188254.1 | 8 | 1468 | ||
NM_001201326.1 | NP_001188255.1 | 6 | 1135 | |||
NM_002614.4 | NP_002605.2 | 9 | 1539 | |||
XM_011509616.2 | XP_011507918.1 | 7 | 1384 | |||
XM_011509618.2 | XP_011507920.1 | 8 | 1472 | |||
XM_011509619.2 | XP_011507921.2 | 9 | 1497 | |||
XM_017001464.1 | XP_016856953.1 | 7 | 1384 | |||
XM_017001465.1 | XP_016856954.1 | 9 | 1497 | |||
XM_017001466.1 | XP_016856955.1 | 9 | 1497 | |||
XM_017001467.1 | XP_016856956.1 | 7 | 1438 | |||
GenBank mRNA | AF012281.1 | - | 8 | 1415 | ||
AK298269.1 | - | 6 | 1076 | |||
AK313100.1 | - | 8 | 1420 | |||
BC006496.2 | - | 8 | 1479 | 2959384 | ||
BC006518.2 | - | 8 | 1479 | 2959384 |
Gene Symbol: PDZK1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111936.1 | 9 | 1512 | |||
GenBank mRNA | AK126719.1 | - | 8 | 2881 |