Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MID1 SIRNA Assays ›
Gene Symbol:
MID1
Gene Name:
midline 1
Gene Aliases:
BBBG1, FXY, GBBB1, MIDIN, OGS1, OS, OSX, RNF59, TRIM18, XPRF, ZNFXY
Chromosome Location:
Chr.X: 10445310 - 11111264 on Build GRCh38
UniGene ID:
Hs.27695
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
118562
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MID1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000381.3
NP_000372.1
2
514
NM_001098624.2
NP_001092094.1
2
444
NM_001193277.1
NP_001180206.1
2
303
NM_001193278.1
NP_001180207.1
1
169
NM_001193279.1
NP_001180208.1
1
169
NM_001193280.1
NP_001180209.1
1
169
NM_001193281.1
NP_001180210.1
1
169
NM_033289.1
NP_150631.1
2
514
NM_033290.3
NP_150632.1
2
306
XM_005274536.1
XP_005274593.1
2
326
XM_005274537.1
XP_005274594.1
2
303
XM_006724492.3
XP_006724555.1
2
925
XM_011545525.2
XP_011543827.1
4
627
XM_011545526.2
XP_011543828.1
4
662
XM_011545527.2
XP_011543829.1
2
572
XM_017029534.1
XP_016885023.1
2
492
XM_017029535.1
XP_016885024.1
4
627
XM_017029536.1
XP_016885025.1
4
742
XM_017029537.1
XP_016885026.1
4
627
XM_017029538.1
XP_016885027.1
2
326
XM_017029539.1
XP_016885028.1
2
488
XM_017029540.1
XP_016885029.1
4
627
XM_017029541.1
XP_016885030.1
4
627
XM_017029542.1
XP_016885031.1
4
627
GenBank mRNA
AF035360.1
-
2
200
AF041206.1
-
2
394
AF041208.1
-
2
514
AF230976.1
-
2
299
AF230977.1
-
2
299
AF269101.1
-
3
422
AK226104.1
-
2
332
AK291215.1
-
2
396
AK297205.1
-
2
460
AK299061.1
-
2
453
AK315095.1
-
2
345
AY539988.1
-
1
113
AY539994.1
-
1
113
AY540022.1
-
1
113
AY540023.1
-
1
113
BC053626.1
-
2
371
6163881
DA435613.1
-
2
303
DA856308.1
-
2
306
DC369426.1
-
2
444
Y13667.1
-
2
299
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
negative regulation of microtubule depolymerization
pattern specification process
positive regulation of stress-activated MAPK cascade
protein localization to microtubule
interferon-gamma-mediated signaling pathway
protein binding
microtubule binding
zinc ion binding
ligase activity
ubiquitin protein ligase binding
identical protein binding
protein homodimerization activity
protein heterodimerization activity
phosphoprotein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-k7c2b:80/10.239.160.46:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0