Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002112.3 | NP_002103.2 | 10 | 1416 | ||
XM_017022094.1 | XP_016877583.1 | 10 | 1285 | |||
XM_017022096.1 | XP_016877585.1 | 9 | 1070 | |||
XM_017022097.1 | XP_016877586.1 | 8 | 1368 | |||
XM_017022098.1 | XP_016877587.1 | 7 | 888 | |||
XM_017022099.1 | XP_016877588.1 | 11 | 1455 | |||
GenBank mRNA | AK027221.1 | - | 3 | 819 | ||
BC130527.1 | - | 10 | 1117 | 40146558 | ||
M60445.1 | - | 10 | 1125 | |||
X54297.1 | - | 10 | 1176 |