Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UXS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253875.1 | NP_001240804.1 | 6, 7 | 577 | ||
NM_025076.4 | NP_079352.2 | 6, 7 | 562 | |||
NR_045607.1 | 5, 6 | 513 | ||||
XM_006712775.3 | XP_006712838.1 | 6, 7 | 539 | |||
XM_011511902.2 | XP_011510204.2 | 6, 7 | 556 | |||
XM_011511903.2 | XP_011510205.2 | 6, 7 | 539 | |||
XM_011511904.2 | XP_011510206.1 | 3, 4 | 884 | |||
XM_017005008.1 | XP_016860497.1 | 6, 7 | 539 | |||
XM_017005009.1 | XP_016860498.1 | 6, 7 | 524 | |||
XM_017005010.1 | XP_016860499.1 | 6, 7 | 524 | |||
XM_017005011.1 | XP_016860500.1 | 6, 7 | 664 | |||
XM_017005012.1 | XP_016860501.1 | 6, 7 | 599 | |||
XM_017005013.1 | XP_016860502.1 | 4, 5 | 1122 | |||
GenBank mRNA | AK075120.1 | - | 6, 7 | 524 | ||
AK075170.1 | - | 6, 7 | 539 | |||
AK122696.1 | - | 5, 6 | 478 | |||
AK290668.1 | - | 6, 7 | 537 | |||
AY147934.1 | - | 6, 7 | 529 | |||
AY358541.1 | - | 6, 7 | 513 | |||
BC009819.2 | - | 6, 7 | 503 | 3843312 | ||
BI825770.1 | - | 6, 7 | 578 | |||
CF242897.1 | - | 6, 7 | 503 |