Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPSB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324081.1 | NP_001311010.1 | 4 | 433 | ||
NM_080861.3 | NP_543137.2 | 4 | 455 | |||
XM_005255673.4 | XP_005255730.1 | 4 | 645 | |||
GenBank mRNA | AF403028.1 | - | 3 | 364 | ||
AK057850.1 | - | 417 | ||||
AK074190.1 | - | 4 | 419 | |||
AK302041.1 | - | 3 | 700 | |||
BC007588.1 | - | 4 | 401 | 3342825 | ||
BC024244.1 | - | 4 | 419 | 4798873 | ||
BC035065.1 | - | 4 | 412 | |||
BC041897.1 | - | 3 | 1229 | 5298729 | ||
BC047441.1 | - | 4 | 397 | 5303706 | ||
BC065515.1 | - | 3 | 540 | |||
BI768095.1 | - | 4 | 409 | |||
BM836743.1 | - | 4 | 480 |