Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048164.2 | NP_001041629.1 | 12 | 1946 | ||
NM_032803.5 | NP_116192.4 | 12 | 1970 | |||
XM_017029912.1 | XP_016885401.1 | 11 | 2461 | |||
GenBank mRNA | AF320612.1 | - | 12 | 1921 | ||
AK027447.1 | - | 10 | 1292 | |||
AK074865.1 | - | 12 | 1941 | |||
AK075014.1 | - | 12 | 1946 | |||
BC033816.1 | - | 12 | 1925 | 5206252 |