Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GABRA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000810.3 | NP_000801.1 | 6 | 951 | ||
NM_001165037.1 | NP_001158509.1 | 6 | 817 | |||
XM_005268258.2 | XP_005268315.1 | 6 | 829 | |||
XM_006720459.2 | XP_006720522.1 | 6 | 733 | |||
XM_017022055.1 | XP_016877544.1 | 6 | 699 | |||
XM_017022056.1 | XP_016877545.1 | 6 | 733 | |||
GenBank mRNA | A28104.1 | - | 6 | 716 | ||
AK290453.1 | - | 6 | 614 | |||
AK294303.1 | - | 3 | 274 | |||
AK303742.1 | - | 1 | 122 | |||
BC111979.1 | - | 5 | 563 | 8327447 | ||
BC113422.1 | - | 5 | 563 | 8322474 | ||
BT009830.1 | - | 4 | 419 | |||
L08485.1 | - | 6 | 724 |