Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000750.3 | NP_000741.1 | 5 | 491 | ||
XM_011521186.2 | XP_011519488.1 | 7 | 521 | |||
XM_011521187.2 | XP_011519489.1 | 6 | 428 | |||
XM_011521190.2 | XP_011519492.1 | 4 | 659 | |||
XM_011521191.2 | XP_011519493.1 | 4 | 239 | |||
XM_011521192.2 | XP_011519494.1 | 5 | 632 | |||
XM_017021885.1 | XP_016877374.1 | 5 | 586 | |||
XM_017021886.1 | XP_016877375.1 | 5 | 2393 | |||
XM_017021887.1 | XP_016877376.1 | 5 | 395 | |||
XM_017021888.1 | XP_016877377.1 | 5 | 395 | |||
XM_017021889.1 | XP_016877378.1 | 5 | 395 | |||
GenBank mRNA | AK310443.1 | - | 12 | 1507 | ||
BC096080.1 | - | 5 | 494 | 40000733 | ||
BC096082.1 | - | 5 | 494 | 40000737 | ||
U48861.1 | - | 5 | 499 | |||
U62439.1 | - | 5 | 465 | |||
X68275.1 | - | 4 | 239 | |||
Y08416.1 | - | 5 | 470 |