Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000792.6 | NP_000783.2 | 4 | 1633 | ||
NM_001039715.2 | NP_001034804.1 | 3 | 1489 | |||
NM_001039716.2 | NP_001034805.1 | 3 | 1433 | |||
NM_001324316.1 | NP_001311245.1 | 2 | 1289 | |||
NM_213593.4 | NP_998758.1 | 4 | 1441 | |||
NR_136692.1 | 4 | 1524 | ||||
NR_136693.1 | 5 | 1550 | ||||
GenBank mRNA | BC017955.1 | - | 3 | 1432 | 4607072 |