Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002372.3 | NP_002363.2 | 3, 4 | 1588 | ||
XM_011543395.2 | XP_011541697.1 | 3, 4 | 1027 | |||
XM_017009472.1 | XP_016864961.1 | 3, 4 | 509 | |||
GenBank mRNA | AK314090.1 | - | 3, 4 | 929 | ||
BC142656.1 | - | 3, 4 | 890 | 40148064 | ||
BC142696.1 | - | 3, 4 | 890 | 40148740 | ||
D63998.1 | - | 3, 4 | 1046 | |||
U31520.1 | - | 3, 4 | 539 |