Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPKC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025194.2 | NP_079470.1 | 1 | 299 | ||
GenBank mRNA | AJ290975.1 | - | 1 | 266 | ||
BC060788.1 | - | 1 | 299 | 30332196 | ||
D38169.1 | - | 1 | 30 |