Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCF2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015078.3 | NP_055893.3 | 4 | 635 | ||
XM_017005943.1 | XP_016861432.1 | 4 | 1356 | |||
XM_017005944.1 | XP_016861433.1 | 4 | 1356 | |||
XM_017005945.1 | XP_016861434.1 | 4 | 1356 | |||
XM_017005946.1 | XP_016861435.1 | 4 | 1356 | |||
GenBank mRNA | AK124500.1 | - | 4 | 467 | ||
AK310962.1 | - | 4 | 609 | |||
AY172737.1 | - | 2 | 187 | |||
BC064632.1 | - | 4 | 451 | 5750039 | ||
BM920156.1 | - | 4 | 439 | |||
BX648525.1 | - | 4 | 411 | |||
BX649003.1 | - | 466 |