Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EYA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005244.4 | NP_005235.3 | 5 | 774 | ||
NM_172110.3 | NP_742108.2 | 5 | 774 | |||
XM_005260327.2 | XP_005260384.1 | 5 | 467 | |||
XM_017027721.1 | XP_016883210.1 | 5 | 492 | |||
GenBank mRNA | AF055015.1 | - | 1 | 88 | ||
AK293032.1 | - | 5 | 478 | |||
AY705349.1 | - | 4 | 410 | |||
BC008803.2 | - | 5 | 456 | 3944547 | ||
BC013882.2 | - | 5 | 774 | |||
BT006682.1 | - | 4 | 328 | |||
U71207.1 | - | 4 | 402 |