Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000480.2 | NP_000471.1 | 5, 6 | 1189 | ||
NM_001025389.1 | NP_001020560.1 | 5, 6 | 1152 | |||
NM_001025390.1 | NP_001020561.1 | 5, 6 | 1291 | |||
NM_001172430.1 | NP_001165901.1 | 5, 6 | 1116 | |||
NM_001172431.1 | NP_001165902.1 | 4, 5 | 812 | |||
GenBank mRNA | AK294444.1 | - | 3, 4 | 514 | ||
AK295046.1 | - | 4, 5 | 810 | |||
AK299113.1 | - | 5, 6 | 864 | |||
AK301507.1 | - | 5, 6 | 1039 | |||
AK302970.1 | - | 5, 6 | 1107 | |||
AK311607.1 | - | 5, 6 | 1212 | |||
AK311711.1 | - | 5, 6 | 1176 | |||
AK313039.1 | - | 5, 6 | 837 | |||
BC126118.1 | - | 5, 6 | 1184 | 8991834 | ||
D12775.1 | - | 5, 6 | 1008 | |||
M84721.1 | - | 5, 6 | 951 | |||
M84722.1 | - | 5, 6 | 1075 |