Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DCTD SIRNA Assays ›
Gene Symbol:
DCTD
Gene Name:
dCMP deaminase
Gene Aliases:
-
Chromosome Location:
Chr.4: 182890091 - 182917477 on Build GRCh38
UniGene ID:
Hs.183850
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
119611
Validated
Purification
HPLC
Standard
In-Vivo Ready
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DCTD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001012732.1
NP_001012750.1
4, 5
562
NM_001921.2
NP_001912.2
4, 5
539
XM_005262778.3
XP_005262835.1
4, 5
587
XM_005262779.3
XP_005262836.1
5, 6
678
XM_005262780.3
XP_005262837.1
6, 7
686
XM_005262781.4
XP_005262838.1
3, 4
344
XM_005262782.3
XP_005262839.1
3, 4
341
XM_006714115.3
XP_006714178.1
4, 5
485
XM_006714116.3
XP_006714179.1
3, 4
782
XM_011531674.2
XP_011529976.1
5, 6
1172
XM_011531675.2
XP_011529977.1
4, 5
903
XM_011531676.2
XP_011529978.1
5, 6
911
XM_017007816.1
XP_016863305.1
5, 6
654
XM_017007817.1
XP_016863306.1
5, 6
621
XM_017007818.1
XP_016863307.1
6, 7
686
XM_017007819.1
XP_016863308.1
6, 7
1180
XM_017007820.1
XP_016863309.1
6, 7
1043
XM_017007821.1
XP_016863310.1
6, 7
1176
XM_017007822.1
XP_016863311.1
6, 7
521
XM_017007823.1
XP_016863312.1
3, 4
632
XM_017007824.1
XP_016863313.1
3, 4
259
GenBank mRNA
AK293135.1
-
3, 4
318
AK313221.1
-
4, 5
464
BC001286.1
-
4, 5
414
3458999
BC088357.1
-
4, 5
453
30389169
BU188090.1
-
3, 4
368
L12136.1
-
3, 4
365
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00157241_m1
Panther Classification:
Molecular Function -
deaminase
hydrolase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
pyrimidine nucleotide metabolic process
nucleotide biosynthetic process
pyrimidine nucleoside biosynthetic process
dCMP deaminase activity
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qskjf:80/10.239.165.3:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0