Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242506.2 | NP_001229435.1 | 15 | 1815 | ||
NM_001242507.2 | NP_001229436.1 | 14 | 1681 | |||
NM_004293.4 | NP_004284.1 | 14 | 1774 | |||
XM_011519217.1 | XP_011517519.1 | 15 | 1886 | |||
XM_017015339.1 | XP_016870828.1 | 14 | 1770 | |||
GenBank mRNA | AB033084.1 | - | 14 | 1641 |