Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SLC26A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000441.1 | NP_000432.1 | 21 | 3194 | ||
XM_005250425.2 | XP_005250482.1 | 21 | 3177 | |||
XM_017012318.1 | XP_016867807.1 | 19 | 2895 | |||
GenBank mRNA | AF030880.1 | - | 21 | 3194 |