Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006841.5 | NP_006832.1 | 16 | 1885 | ||
XM_006712954.2 | XP_006713017.1 | 16 | 1919 | |||
GenBank mRNA | AF244548.1 | - | 16 | 1844 | ||
BC042875.1 | - | 16 | 1857 | 5174617 | ||
BX537382.1 | - | 16 | 1859 | |||
U49082.1 | - | 16 | 1855 |