Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321036.1 | NP_001307965.1 | 2 | 536 | ||
NM_013309.5 | NP_037441.2 | 2 | 536 | |||
XM_011521997.2 | XP_011520299.1 | 2 | 1040 | |||
XM_017022560.1 | XP_016878049.1 | 1 | 342 | |||
XM_017022561.1 | XP_016878050.1 | 2 | 534 | |||
XM_017022562.1 | XP_016878051.1 | 1 | 342 | |||
GenBank mRNA | AF025409.1 | - | 1 | 224 | ||
AK290874.1 | - | 1 | 342 | |||
BC026089.2 | - | 2 | 536 | 4829839 | ||
BM789500.1 | - | 1 | 106 |