Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258426.1 | NP_001245355.1 | 14 | 1985 | ||
NM_001258427.1 | NP_001245356.1 | 13 | 1838 | |||
NM_001258428.1 | NP_001245357.1 | 14 | 2099 | |||
NM_031467.2 | NP_113655.2 | 14 | 2027 | |||
XM_005268521.3 | XP_005268578.2 | 14 | 2584 | |||
XM_011537694.2 | XP_011535996.1 | 14 | 2656 | |||
XM_017009933.1 | XP_016865422.1 | 14 | 2656 | |||
XM_017009934.1 | XP_016865423.1 | 14 | 2647 | |||
XM_017009935.1 | XP_016865424.1 | 14 | 2623 | |||
XM_017009936.1 | XP_016865425.1 | 14 | 2584 | |||
XM_017009937.1 | XP_016865426.1 | 13 | 2542 | |||
XM_017009938.1 | XP_016865427.1 | 14 | 2491 | |||
XM_017009939.1 | XP_016865428.1 | 14 | 2542 | |||
XM_017009940.1 | XP_016865429.1 | 13 | 2467 | |||
XM_017009941.1 | XP_016865430.1 | 13 | 2458 | |||
XM_017009942.1 | XP_016865431.1 | 12 | 2347 | |||
XM_017009943.1 | XP_016865432.1 | 11 | 1658 | |||
GenBank mRNA | AB032762.1 | - | 14 | 1986 | ||
AF313465.1 | - | 14 | 2086 | |||
AF332961.1 | - | 14 | 2027 | |||
AF336237.1 | - | 14 | 1977 | |||
BC136262.1 | - | 13 | 1829 | 9020249 | ||
BC143602.1 | - | 13 | 1829 | 9052115 |