Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000714.5 | NP_000705.2 | 4 | 571 | ||
NM_001256530.1 | NP_001243459.1 | 4 | 766 | |||
NM_001256531.1 | NP_001243460.1 | 4 | 675 | |||
NR_046308.1 | 3 | 360 | ||||
GenBank mRNA | AF075589.1 | - | 3 | 374 | ||
AF075590.1 | - | 3 | 374 | |||
AY998017.1 | - | 3 | 466 | |||
BC001110.2 | - | 4 | 521 | 2989070 | ||
BG749252.1 | - | 4 | 675 | |||
BG752086.1 | - | 4 | 572 | |||
BQ057964.1 | - | 556 | ||||
BT006949.1 | - | 3 | 451 | |||
CR456409.1 | - | 3 | 460 | |||
CR457007.1 | - | 2 | 170 | |||
L21950.1 | - | 3 | 330 | |||
M36035.1 | - | 4 | 512 |