Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FOLR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000803.4 | NP_000794.3 | 4, 5 | 653 | ||
NM_001113534.1 | NP_001107006.1 | 4, 5 | 647 | |||
NM_001113535.1 | NP_001107007.1 | 4, 5 | 641 | |||
NM_001113536.1 | NP_001107008.1 | 4, 5 | 635 | |||
XM_005273856.3 | XP_005273913.1 | 4, 5 | 651 | |||
GenBank mRNA | AK222539.1 | - | 4, 5 | 644 | ||
AK222643.1 | - | 4, 5 | 635 | |||
BC027894.1 | - | 4, 5 | 622 | |||
BC058036.1 | - | 4, 5 | 637 | 6662871 | ||
BC115366.1 | - | 3, 4 | 423 | |||
CD014063.1 | - | 3, 4 | 504 | |||
CD692073.1 | - | 4, 5 | 651 |