Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A18AS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302862.1 | NP_001289791.1 | 3 | 916 | ||
NM_007105.3 | NP_009036.2 | 4 | 1311 | |||
XM_017017832.1 | XP_016873321.1 | 2 | 2349 | |||
XM_017017833.1 | XP_016873322.1 | 4 | 2732 | |||
XM_017017834.1 | XP_016873323.1 | 2 | 2013 | |||
XM_017017835.1 | XP_016873324.1 | 2 | 1792 | |||
XM_017017836.1 | XP_016873325.1 | 2 | 657 | |||
GenBank mRNA | AF035407.1 | - | 2 | 678 | ||
AF037066.1 | - | 4 | 1107 | |||
BC030237.1 | - | 3 | 472 | 4185944 |