Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RAB9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195328.1 | NP_001182257.1 | 2 | 637 | ||
NM_004251.4 | NP_004242.1 | 3 | 726 | |||
XM_017029964.1 | XP_016885453.1 | 3 | 1128 | |||
GenBank mRNA | AF498944.1 | - | 1 | 443 | ||
AK290505.1 | - | 2 | 616 | |||
BC017265.2 | - | 3 | 625 | 4139714 | ||
CR450362.1 | - | 1 | 443 | |||
U44103.1 | - | 1 | 443 |