Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243738.1 | NP_001230667.1 | 4, 5 | 842 | ||
NM_004761.4 | NP_004752.1 | 5, 6 | 1039 | |||
XM_011514781.1 | XP_011513083.1 | 3, 4 | 311 | |||
XM_011514785.2 | XP_011513087.1 | 3, 4 | 311 | |||
GenBank mRNA | AB012295.1 | - | 4, 5 | 391 | ||
AK294442.1 | - | 4, 5 | 378 | |||
AL050259.1 | - | 5, 6 | 612 | |||
BC005037.1 | - | 2, 3 | 80 | |||
BC032681.1 | - | 5, 6 | 594 | 5585507 | ||
BX648015.1 | - | 4, 5 | 384 |