Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASGRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128602.1 | NP_001122074.1 | 8 | 1044 | ||
NM_001306086.1 | NP_001293015.1 | 8 | 1044 | |||
NM_005739.3 | NP_005730.2 | 8 | 1044 | |||
XM_005254114.2 | XP_005254171.1 | 8 | 1033 | |||
XM_011521151.2 | XP_011519453.1 | 8 | 1033 | |||
XM_017021860.1 | XP_016877349.1 | 8 | 971 | |||
GenBank mRNA | AB208848.1 | - | 2 | 170 | ||
AF081195.1 | - | 8 | 969 | |||
AF081197.1 | - | 7 | 823 | |||
AF106071.1 | - | 8 | 878 | |||
AK295097.1 | - | 7 | 908 | |||
AK314091.1 | - | 8 | 1044 | |||
AK316067.1 | - | 8 | 972 | |||
AY634315.2 | - | 8 | 873 | |||
AY858556.2 | - | 8 | 873 | |||
AY954625.4 | - | 8 | 873 | |||
AY966005.1 | - | 8 | 873 | |||
BC067298.1 | - | 8 | 949 | |||
BC109296.1 | - | 8 | 916 | 40034427 | ||
BC109297.1 | - | 8 | 916 | 40034428 |