Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289123.1 | NP_001276052.1 | 3, 4 | 454 | ||
NM_001289127.1 | NP_001276056.1 | 3, 4 | 436 | |||
NM_001289129.1 | NP_001276058.1 | 3, 4 | 384 | |||
NM_001289130.1 | NP_001276059.1 | 2, 3 | 493 | |||
NM_001289131.1 | NP_001276060.1 | 2, 3 | 460 | |||
NM_006087.3 | NP_006078.2 | 2, 3 | 536 | |||
GenBank mRNA | AK075307.1 | - | 2, 3 | 262 | ||
AK095202.1 | - | 1, 2 | 793 | |||
AK293503.1 | - | 2, 3 | 185 | |||
AK295917.1 | - | 2, 3 | 261 | |||
BC006570.2 | - | 2, 3 | 245 | 3603726 | ||
BC013683.1 | - | 2, 3 | 271 | 4155617 | ||
DA265791.1 | - | 2, 3 | 221 | |||
DA302559.1 | - | 2, 3 | 214 | |||
DC303150.1 | - | 3, 4 | 409 | |||
DC314277.1 | - | 2, 3 | 536 | |||
DC353527.1 | - | 3, 4 | 384 | |||
DC354036.1 | - | 3, 4 | 387 |