Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289123.1 | NP_001276052.1 | 5 | 1817 | ||
NM_001289127.1 | NP_001276056.1 | 5 | 1799 | |||
NM_001289129.1 | NP_001276058.1 | 5 | 1747 | |||
NM_001289130.1 | NP_001276059.1 | 4 | 1856 | |||
NM_001289131.1 | NP_001276060.1 | 4 | 1823 | |||
NM_006087.3 | NP_006078.2 | 4 | 1899 | |||
GenBank mRNA | AK075307.1 | - | 4 | 1625 | ||
AK294102.1 | - | 3 | 1000 | |||
AK295917.1 | - | 5 | 1378 | |||
BC006570.2 | - | 4 | 1608 | 3603726 | ||
BC013683.1 | - | 4 | 1634 | 4155617 |