Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASL10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006477.4 | NP_006468.1 | 3 | 1325 | ||
XM_011529821.1 | XP_011528123.1 | 4 | 1043 | |||
XM_011529822.1 | XP_011528124.1 | 4 | 1799 | |||
XM_011529823.1 | XP_011528125.1 | 3 | 906 | |||
GenBank mRNA | BC022473.1 | - | 3 | 1325 | 4794714 | |
BC040434.1 | - | 2 | 1349 | 5727949 | ||
BC050739.1 | - | 2 | 1555 | 5244128 | ||
BC058077.1 | - | 3 | 1234 | 5240551 | ||
BX111994.1 | - | 1 | 372 | |||
Y07847.1 | - | 3 | 1136 |