Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281741.1 | NP_001268670.1 | 5 | 493 | ||
NM_001281742.1 | NP_001268671.1 | 5 | 429 | |||
NM_007019.3 | NP_008950.1 | 6 | 634 | |||
NM_181799.2 | NP_861515.1 | 6 | 580 | |||
NM_181800.2 | NP_861516.1 | 5 | 547 | |||
NM_181801.3 | NP_861517.1 | 6 | 671 | |||
NR_104036.1 | 4 | 342 | ||||
NR_104037.1 | 5 | 437 | ||||
GenBank mRNA | AK311949.1 | - | 6 | 601 | ||
BC007656.2 | - | 6 | 557 | 3345575 | ||
BC016292.1 | - | 6 | 563 | 3910286 | ||
BC028161.1 | - | 5 | 1173 | |||
BC050736.1 | - | 6 | 584 | 6503915 | ||
BI858659.1 | - | 6 | 671 | |||
BM556795.1 | - | 6 | 498 | |||
BT007300.1 | - | 6 | 514 | |||
BT020165.1 | - | 6 | 514 | |||
BT020166.1 | - | 6 | 514 | |||
BU169315.1 | - | 6 | 521 | |||
BU171488.1 | - | 5 | 453 | |||
BU844974.1 | - | 5 | 449 | |||
CB143441.1 | - | 4 | 260 | |||
CK904708.1 | - | 5 | 459 | |||
CN354813.1 | - | 5 | 330 | |||
CN354815.1 | - | 5 | 372 | |||
CR456988.1 | - | 6 | 514 | |||
U73379.1 | - | 6 | 554 |