Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024539.3 | NP_078815.3 | 2, 3 | 829 | ||
NM_194463.1 | NP_919445.1 | 2, 3 | 992 | |||
GenBank mRNA | AF394689.1 | - | 2, 3 | 752 | ||
AK074264.1 | - | 2, 3 | 992 | |||
AK126553.1 | - | 2, 3 | 812 | |||
BC030951.1 | - | 1, 2 | 105 | |||
BC036901.1 | - | 2, 3 | 841 | |||
BC063404.1 | - | 2, 3 | 831 | 5191785 |