Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCL2L12 SIRNA Assays ›
Gene Symbol:
BCL2L12
Gene Name:
BCL2 like 12
Gene Aliases:
-
Chromosome Location:
Chr.19: 49665072 - 49673916 on Build GRCh38
UniGene ID:
Hs.289052
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
120838
Validated
Purification
HPLC
Standard
In-Vivo Ready
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCL2L12
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001040668.1
NP_001035758.1
7
1759
NM_001282516.1
NP_001269445.1
6
1489
NM_001282517.1
NP_001269446.1
5
1346
NM_001282519.1
NP_001269448.1
6
1538
NM_001282520.1
NP_001269449.1
5
1397
NM_001282521.1
NP_001269450.1
4
1310
NM_138639.1
NP_619580.1
7
1762
NR_104200.1
6
1619
NR_104201.1
7
1811
NR_104202.1
6
1668
NR_104203.1
5
1395
NR_104204.1
4
1254
NR_104205.1
3
1167
XM_017027345.1
XP_016882834.1
6
1556
GenBank mRNA
BC007724.2
-
7
897
BC104004.1
-
7
1335
40024338
BC104005.1
-
7
1338
40024342
BC104006.1
-
7
1338
40024345
KR820244.1
-
3
1551
KR820245.1
-
5
1916
KR820246.1
-
4
1643
KR820247.1
-
4
1692
KR820248.1
-
5
1965
KR820249.1
-
3
1600
KR820250.1
-
5
1965
KR820251.1
-
4
1692
KR820252.1
-
4
1741
KR820253.1
-
5
2014
KR820254.1
-
4
1332
KR820255.1
-
5
1424
KR820256.1
-
5
1473
KR820257.1
-
6
1746
KR820258.1
-
4
1381
KR820259.1
-
6
1746
KR820260.1
-
5
1473
KR820261.1
-
5
1522
KR820262.1
-
6
1795
KR820263.1
-
4
1329
KR820264.1
-
5
1421
KR820265.1
-
5
1470
KR820266.1
-
4
1378
KR820267.1
-
6
1743
KR820268.1
-
5
1470
KR820269.1
-
5
1519
KR820270.1
-
4
934
KR820271.1
-
6
1299
KR820272.1
-
5
1026
KR820273.1
-
5
1075
KR820274.1
-
4
983
KR820275.1
-
6
1348
KR820276.1
-
5
1075
KR820277.1
-
5
1124
KR820278.1
-
5
764
KR820279.1
-
7
1129
KR820280.1
-
6
856
KR820281.1
-
6
905
KR820282.1
-
6
804
KR820283.1
-
6
853
KR820284.1
-
5
761
KR820285.1
-
7
1126
KR820286.1
-
6
853
KR820287.1
-
6
902
KR820288.1
-
3
1189
KR820289.1
-
4
1281
KR820290.1
-
4
1330
KR820291.1
-
5
1554
KR820292.1
-
5
1603
KR820293.1
-
2
1102
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00369327_m1
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
positive regulation of transcription from RNA polymerase II promoter
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
inhibition of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of cellular senescence
p53 binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7cf5v:80/10.239.163.60:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0