Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002703.4 | NP_002694.3 | 11 | 2474 | ||
XM_011534387.1 | XP_011532689.1 | 11 | 2251 | |||
GenBank mRNA | AB209246.1 | - | 3 | 2818 | ||
U00238.1 | - | 11 | 2366 |