Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RABL2B SIRNA Assays ›
Gene Symbol:
RABL2B
RABL2A
Gene Name:
RAB, member of RAS oncogene family-like 2B
RAB, member of RAS oncogene family-like 2A
Gene Aliases:
-
-
Chromosome Location:
Chr.22: 50767492 - 50783705 on Build GRCh38
Chromosome Location:
Chr.2: 113627218 - 113643398 on Build GRCh38
UniGene ID:
Hs.584862
Hs.446425
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
120918
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RABL2B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001003789.1
NP_001003789.1
4, 5
427
NM_001130919.1
NP_001124391.1
4, 5
423
NM_001130920.1
NP_001124392.1
5, 6
582
NM_001130921.1
NP_001124393.1
5, 6
675
NM_001130922.1
NP_001124394.1
4, 5
427
NM_001130923.1
NP_001124395.1
5, 6
582
NM_007081.2
NP_009012.1
4, 5
423
XM_005261899.2
XP_005261956.1
5, 6
679
XM_011530668.1
XP_011528970.1
5, 6
679
XM_011530669.2
XP_011528971.1
4, 5
426
XM_011530670.1
XP_011528972.1
4, 5
443
XM_011530671.1
XP_011528973.1
5, 6
553
XM_011530673.1
XP_011528975.1
5, 6
679
XM_011530675.1
XP_011528977.1
5, 6
679
XM_017028543.1
XP_016884032.1
5, 6
869
XM_017028544.1
XP_016884033.1
5, 6
774
XM_017028545.1
XP_016884034.1
5, 6
715
XM_017028546.1
XP_016884035.1
5, 6
717
XM_017028547.1
XP_016884036.1
5, 6
869
XM_017028548.1
XP_016884037.1
4, 5
426
XM_017028549.1
XP_016884038.1
5, 6
715
XM_017028550.1
XP_016884039.1
5, 6
553
XM_017028551.1
XP_016884040.1
3, 4
600
XM_017028552.1
XP_016884041.1
5, 6
869
XM_017028553.1
XP_016884042.1
5, 6
771
XM_017028554.1
XP_016884043.1
5, 6
715
XM_017028555.1
XP_016884044.1
5, 6
717
XM_017028556.1
XP_016884045.1
5, 6
869
XM_017028557.1
XP_016884046.1
5, 6
715
XM_017028558.1
XP_016884047.1
5, 6
553
XM_017028559.1
XP_016884048.1
3, 4
600
GenBank mRNA
AF095352.1
-
4, 5
364
AK308268.1
-
4, 5
400
AK313442.1
-
5, 6
679
BC014879.2
-
4, 5
389
3906749
BC024281.1
-
5, 6
625
4580599
BG177165.1
-
423
BT020037.1
-
3, 4
211
BX421088.2
-
5, 6
528
BX432293.2
-
4, 5
369
Gene Symbol: RABL2A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
CN342140.1
-
3, 4
268
DB451301.2
-
4, 5
391
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
GTPase activity
GTP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0