Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000134.3 | NP_000125.2 | 3 | 353 | ||
GenBank mRNA | AK025079.1 | - | 3 | 325 | ||
BC069466.1 | - | 3 | 344 | 7262448 | ||
BC069617.1 | - | 3 | 337 | 7262424 | ||
BC069625.1 | - | 3 | 344 | 7262436 | ||
BC069637.1 | - | 3 | 344 | 7262460 | ||
BC111791.1 | - | 3 | 353 | 40017872 |