Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004139.4 | NP_004130.2 | 7, 8 | 846 | ||
GenBank mRNA | AF105067.1 | - | 7, 8 | 757 | ||
AK313625.1 | - | 7, 8 | 776 | |||
BC022256.1 | - | 7, 8 | 775 | 4767724 | ||
M35533.1 | - | 7, 8 | 740 |