Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004164.2 | NP_004155.2 | 2 | 178 | ||
GenBank mRNA | AK291978.1 | - | 2 | 184 | ||
BC069296.1 | - | 2 | 127 | 7262683 | ||
BC069361.1 | - | 2 | 127 | 7262695 | ||
BC069396.1 | - | 2 | 127 | 7262707 | ||
BC069424.1 | - | 2 | 127 | 7262719 | ||
BC069447.1 | - | 2 | 127 | 7262731 | ||
BC069469.1 | - | 2 | 126 | |||
BC069513.1 | - | 2 | 127 | 7262755 | ||
BC069522.1 | - | 2 | 127 | 7262672 | ||
BX282005.1 | - | 4 | 305 | |||
U13831.1 | - | 2 | 127 |