Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304485.1 | NP_001291414.1 | 5 | 610 | ||
NM_001304486.1 | NP_001291415.1 | 4 | 488 | |||
NM_001304487.1 | NP_001291416.1 | 4 | 522 | |||
NM_001304488.1 | NP_001291417.1 | 5 | 507 | |||
NM_001304489.1 | NP_001291418.1 | 5 | 492 | |||
NM_001304490.1 | NP_001291419.1 | 5 | 489 | |||
NM_001304491.1 | NP_001291420.1 | 4 | 401 | |||
NM_012460.3 | NP_036592.1 | 6 | 799 | |||
NR_130750.1 | 5 | 734 | ||||
NR_130751.1 | 4 | 424 | ||||
NR_130752.1 | 3 | 423 | ||||
NR_130753.1 | 3 | 415 | ||||
GenBank mRNA | AF150100.1 | - | 5 | 490 | ||
AK000666.1 | - | 5 | 508 | |||
BC020213.1 | - | 6 | 699 | 4640518 | ||
BC054875.1 | - | 4 | 412 | 6663143 | ||
BF034731.1 | - | 5 | 454 | |||
BG249379.1 | - | 5 | 379 | |||
BM811095.1 | - | 5 | 646 | |||
DB110327.1 | - | 3 | 352 | |||
DN912572.1 | - | 5 | 421 | |||
HY041857.1 | - | 4 | 304 | |||
HY096567.1 | - | 4 | 353 | |||
W74224.1 | - | 5 | 395 |