Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GLYR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308096.1 | NP_001295025.1 | 4 | 277 | ||
NM_001324096.1 | NP_001311025.1 | 4 | 363 | |||
NM_001324097.1 | NP_001311026.1 | 4 | 363 | |||
NM_001324098.1 | NP_001311027.1 | 4 | 363 | |||
NM_032569.3 | NP_115958.2 | 4 | 277 | |||
NR_136695.1 | 5 | 562 | ||||
NR_136696.1 | 4 | 363 | ||||
NR_136697.1 | 4 | 363 | ||||
NR_136698.1 | 4 | 363 | ||||
NR_136699.1 | 4 | 363 | ||||
NR_136700.1 | 4 | 363 | ||||
XM_005255639.4 | XP_005255696.2 | 4 | 321 | |||
XM_011522716.2 | XP_011521018.1 | 4 | 405 | |||
XM_011522717.1 | XP_011521019.1 | 5 | 462 | |||
XM_017023786.1 | XP_016879275.1 | 4 | 358 | |||
XM_017023787.1 | XP_016879276.1 | 4 | 358 | |||
XM_017023788.1 | XP_016879277.1 | 5 | 462 | |||
XM_017023789.1 | XP_016879278.1 | 5 | 462 | |||
GenBank mRNA | AF244907.1 | - | 4 | 258 | ||
AF326966.1 | - | 4 | 240 | |||
AK296842.1 | - | 4 | 254 | |||
AK307834.1 | - | 4 | 253 | |||
AY352585.1 | - | 4 | 240 | |||
BC003693.1 | - | 3 | 202 | |||
BC032855.1 | - | 4 | 268 | 5265561 | ||
BC047223.1 | - | 4 | 257 | 5263991 | ||
BC064940.1 | - | 5 | 461 | 5787345 | ||
DB492328.1 | - | 4 | 282 | |||
HY130375.1 | - | 4 | 364 |