Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SFXN5 SIRNA Assays ›
Gene Symbol:
SFXN5
Gene Name:
sideroflexin 5
Gene Aliases:
BBG-TCC
Chromosome Location:
Chr.2: 72942036 - 73072234 on Build GRCh38
UniGene ID:
Hs.368171
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
121108
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SFXN5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_144579.2
NP_653180.1
4, 5
412
XM_005264648.1
XP_005264705.1
4, 5
318
XM_006712147.1
XP_006712210.1
4, 5
318
XM_011533186.2
XP_011531488.1
5, 6
1960
XM_011533187.2
XP_011531489.1
5, 6
1957
XM_011533188.2
XP_011531490.1
4, 5
1815
XM_011533189.1
XP_011531491.1
4, 5
318
XM_011533190.2
XP_011531492.1
5, 6
865
XM_011533192.2
XP_011531494.1
5, 6
1099
XM_011533194.2
XP_011531496.1
5, 6
1958
XM_011533195.2
XP_011531497.1
5, 6
1960
XM_011533196.2
XP_011531498.1
4, 5
1044
XM_011533197.1
XP_011531499.1
4, 5
318
XM_017005340.1
XP_016860829.1
4, 5
1812
XM_017005341.1
XP_016860830.1
4, 5
1813
XM_017005342.1
XP_016860831.1
4, 5
318
XM_017005343.1
XP_016860832.1
5, 6
1099
XM_017005344.1
XP_016860833.1
4, 5
1044
GenBank mRNA
AK024193.1
-
3, 4
940
AK289731.1
-
4, 5
301
AK294751.1
-
4, 5
349
AK295637.1
-
4, 5
223
AK314770.1
-
4, 5
300
AY044437.1
-
4, 5
416
BC013233.1
-
4, 5
353
BC101311.2
-
4, 5
412
40024811
BC101312.1
-
4, 5
412
40024812
BC101313.2
-
4, 5
412
40024813
Back To Top
More Information
Panther Classification:
Molecular Function -
primary active transporter
Gene Ontology Categories:
Function(s)
Process(es)
citrate transport
ion transmembrane transport
iron ion homeostasis
citrate transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-462rt:80/10.239.167.23:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0