Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015306.2 | NP_056121.2 | 62 | 7597 | ||
XM_005270690.3 | XP_005270747.1 | 49 | 5638 | |||
XM_017000831.1 | XP_016856320.1 | 63 | 7662 | |||
XM_017000832.1 | XP_016856321.1 | 63 | 7659 | |||
XM_017000833.1 | XP_016856322.1 | 63 | 7154 | |||
XM_017000834.1 | XP_016856323.1 | 62 | 7094 | |||
GenBank mRNA | AB028980.1 | - | 21 | 2416 | ||
AK000321.1 | - | 6 | 581 | |||
AL832370.1 | - | 24 | 2823 | |||
BC029660.1 | - | 7 | 650 |