Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303058.1 | NP_001289987.1 | 2 | 274 | ||
NM_001303059.1 | NP_001289988.1 | 2 | 274 | |||
NM_001303060.1 | NP_001289989.1 | 1 | 497 | |||
NM_001303061.1 | NP_001289990.1 | 2 | 267 | |||
NM_001303062.1 | NP_001289991.1 | 1 | 497 | |||
NM_182547.3 | NP_872353.2 | 2 | 274 | |||
GenBank mRNA | AK074557.1 | - | 2 | 210 | ||
AK290125.1 | - | 2 | 220 | |||
AL602839.1 | - | 2 | 181 | |||
AY191223.1 | - | 2 | 193 | |||
BC035467.1 | - | 2 | 197 | |||
BC052641.1 | - | 2 | 207 | 5223093 | ||
BC053583.1 | - | 2 | 207 | |||
BC057851.1 | - | 2 | 210 | 4798030 | ||
BP872004.1 | - | 2 | 266 | |||
BX461603.2 | - | 1 | 396 | |||
BX647965.1 | - | 2 | 190 | |||
DA419871.1 | - | 2 | 274 |