Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G2F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022819.3 | NP_073730.3 | 4 | 501 | ||
XM_011541955.1 | XP_011540257.1 | 4 | 494 | |||
XM_011541956.1 | XP_011540258.1 | 4 | 505 | |||
GenBank mRNA | AF306566.1 | - | 3 | 274 | ||
AK093645.1 | - | 4 | 505 |