Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282232.1 | NP_001269161.1 | 5 | 666 | ||
NM_001282233.1 | NP_001269162.1 | 4 | 523 | |||
NM_001282234.1 | NP_001269163.1 | 5 | 543 | |||
NM_002791.2 | NP_002782.1 | 5 | 618 | |||
XM_017021468.1 | XP_016876957.1 | 5 | 1954 | |||
GenBank mRNA | AK298920.1 | - | 4 | 491 | ||
AK302008.1 | - | 5 | 468 | |||
AK313011.1 | - | 5 | 544 | |||
AK316223.1 | - | 5 | 532 | |||
BC002979.1 | - | 5 | 509 | 3542966 | ||
BC017882.1 | - | 5 | 531 | 4278372 | ||
BC022354.1 | - | 5 | 488 | 4292526 | ||
BC023659.1 | - | 5 | 536 | 4869452 | ||
BC070137.1 | - | 5 | 540 | 6458246 | ||
BG701535.1 | - | 5 | 599 | |||
CR456944.1 | - | 5 | 458 | |||
X59417.1 | - | 5 | 520 | |||
X61972.1 | - | 4 | 383 |