Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001273.3 | NP_001264.2 | 3, 4 | 439 | ||
XM_006718958.1 | XP_006719021.1 | 3, 4 | 445 | |||
XM_006718959.1 | XP_006719022.1 | 3, 4 | 445 | |||
XM_006718960.1 | XP_006719023.1 | 3, 4 | 445 | |||
XM_006718962.1 | XP_006719025.1 | 3, 4 | 445 | |||
XM_017018725.1 | XP_016874214.1 | 3, 4 | 445 | |||
XM_017018726.1 | XP_016874215.1 | 3, 4 | 445 | |||
XM_017018727.1 | XP_016874216.1 | 3, 4 | 445 | |||
XM_017018728.1 | XP_016874217.1 | 3, 4 | 445 | |||
XM_017018730.1 | XP_016874219.1 | 3, 4 | 445 | |||
XM_017018731.1 | XP_016874220.1 | 3, 4 | 445 | |||
GenBank mRNA | BC014964.1 | - | 3, 4 | 365 | ||
BC025962.1 | - | 3, 4 | 365 | |||
BC038596.1 | - | 3, 4 | 380 | 5528023 | ||
X86691.1 | - | 2, 3 | 316 |