Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002803.3 | NP_002794.1 | 12 | 1507 | ||
GenBank mRNA | AB075520.1 | - | 12 | 1368 | ||
AK298821.1 | - | 11 | 1268 | |||
BC002589.2 | - | 12 | 1351 | 3161790 | ||
D11094.1 | - | 12 | 1385 |