Gene Symbol: PARN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134477.2 | NP_001127949.1 | 5 | 357 | ||
NM_002582.3 | NP_002573.1 | 5 | 440 | |||
XM_011522510.2 | XP_011520812.1 | 5 | 446 | |||
XM_011522511.2 | XP_011520813.1 | 5 | 445 | |||
XM_011522513.2 | XP_011520815.1 | 5 | 356 | |||
XM_011522514.1 | XP_011520816.1 | 5 | 444 | |||
XM_017023258.1 | XP_016878747.1 | 5 | 444 | |||
XM_017023259.1 | XP_016878748.1 | 2 | 89 | |||
XM_017023260.1 | XP_016878749.1 | 1 | 39 | |||
GenBank mRNA | AJ005698.1 | - | 5 | 351 | ||
AK293189.1 | - | 5 | 349 | |||
AK315020.1 | - | 5 | 436 | |||
BC044763.1 | - | 5 | 378 | |||
BC050029.1 | - | 5 | 378 | 4825218 | ||
DA308295.1 | - | 5 | 442 |